Human Recombinant CD137/4-1BB Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
10041-H02H-1 10041-H02H-100
103885-932EA 1957.38 USD InStock
103885-932 103885-970
Human Recombinant CD137/4-1BB Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human TNFRSF9 (NP_001552.2) (Met1-Gln186) was expressed with the Fc region of human IgG1 at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty