Human Recombinant IL12A/NKSF1 Protein (His Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
10021-H08H-100 10021-H08H-1
103620-726EA 371.68 USD InStock
103620-726 103620-728 103678-232 103678-946 103678-948
Human Recombinant IL12A/NKSF1 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the p35 subunit of human IL12, termed as IL12A (P29459) (Met 1-Ser 219) was expressed, fused with a polyhistidine tag at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty