Human Recombinant TNF-beta/TNFSF1/Lymphotoxin alpha Protein (from E. coli)
Supplier: wmf consumer eletric GmbH
10270-HNAE-100 10270-HNAE-1
103622-372EA 351.48 USD InStock
103622-372 103677-458
Human Recombinant TNF-beta/TNFSF1/Lymphotoxin alpha Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human TNFβ (P01374) (Leu 35-Leu 205) was expressed and purified, with an initial Met at the N-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty