Influenza A H1N1 (A/California/04/2009) Recombinant Neuraminidase/NA (Active) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
11058-VNAHC-100 11058-VNAHC-200
103633-848EA 171.7 USD InStock
103633-848 103633-850
Influenza A H1N1 (A/California/04/2009) Recombinant Neuraminidase/NA (Active) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the Influenza A virus (A/California/04/2009 (H1N1)) neuraminidase (ACP41107.1) (His 36-Lys 469) was expressed, the cell lysates are collected, and bio-activity was tested.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty