Mouse Recombinant IL-1 alpha/IL1A/IL1F1 Protein (from E. coli)
Supplier: wmf consumer eletric GmbH
50114-MNAE-100 50114-MNAE-20
103664-610EA 452.48 USD InStock
103664-610 103666-794 103677-926
Mouse Recombinant IL-1 alpha/IL1A/IL1F1 Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of mouse IL1A (NP_034684.2) (Ser 115-Ser 270) was expressed, with an initial Met at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty