Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ProLite™ Orange 5000X stain_protein gel

Supplier: AAT BIOQUEST INC
Warning

18000 18001
76485-810EA 112.97 USD InStock
76485-810 76485-812
ProLite™ Orange 5000X stain_protein gel
ProLite™ Orange
Prolite™ Orange is an alternative protein stain that can be used to replace SYPRO Orange Protein Gel Stain (SYPRO is the trademark of ThermoFisher).

  • Superior sensitivity
  • Compataible with all major viewing platforms
  • Rapid staining
  • Can be used for total protein quantitation
  • Alternative to sypro orange

Precaución: For research use only


MW: 486.72 g/mol
ADR: 5 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty