Fluo-8® potassium salt
Supplier: AAT BIOQUEST INC
Warning
21087 21089
76483-238EA 375.27 USD InStock
76483-238 76483-240
Fluo-8® potassium salt
Fluo-8® potassium salt
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms
Precaución: For research use only
ADR: 5,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30c8634a5ce98ab3181aa7e99e362393159f01b2&vid=30c8634a5ce98ab3181aa7e99e362393159f01b2&prid=web&clid=SAPDAM
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty