Brady® Standard Key Retaining Lockout Aluminum Padlocks with Steel Shackle, Brady
Supplier: Brady Worldwide
SDAL-RED-38ST-KD SDAL-YLW-38ST-KD SDAL-BLU-38ST-KD SDAL-ORG-38ST-KD SDAL-GRN-38ST-KD SDAL-RED-38ST-KD6 SDAL-YLW-38ST-KD6 SDAL-BLU-38ST-KD6 SDAL-ORG-38ST-KD6 SDAL-GRN-38ST-KD6 SDAL-RED-38ST-KA6 SDAL-YLW-38ST-KA6 SDAL-BLU-38ST-KA6 SDAL-GRN-38ST-KA6 SDAL-ORG-38ST-KA6 SDAL-RED-38ST-KD12 SDAL-RED-38ST-KA12 SDAL-RED-38ST-KD24 SDAL-RED-38ST-KA24
76537-246EA 14.42 USD InStock
76537-246 76537-248 76537-250 76537-252 76537-254 76537-256 76537-258 76537-260 76537-262 76537-264 76537-266 76537-268 76537-270 76537-272 76537-274 76537-276 76537-278 76537-280 76537-282
Brady® Standard Key Retaining Lockout Aluminum Padlocks with Steel Shackle, Brady
Security Locks
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient, and cost-effective Safety Lockout Padlocks from Brady.
  • Standard padlock body type
  • Service temperature range: −15 to +250 °F
Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program. Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
Aluminum padlocks with steel shackles are durable, corrosion-resistant, and ideal for general lockoutMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty