Brady® Repositionable Multi-Purpose Vinyl Labels with Ribbon, Brady
Supplier: Brady Worldwide
M21-500-581-WT M21-750-581-WT
76537-140RL 27.61 USD InStock
76537-140 76537-142
Brady® Repositionable Multi-Purpose Vinyl Labels with Ribbon, Brady
Labels
Repositionable Vinyl Film (B-581) is a durable, flexible, high-gloss material ideal for indoor/outdoor marking where frequent repositioning is required.
  • Repositionable acrylic adhesive
  • Gloss finish
  • Repositionable design
Special aggressive adhesive stays in place, but removes cleanly without leaving adhesive residue, making it ideal for applications like 5S and inventory. Vinyl film can be thermal transfer printed. Average outdoor durability is 5 years. Service temperature of −40 to +180° F (−40 to +82° C).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty