Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium thiosulphate pentahydrate 99.5+% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC42446-0010 AC42446-0051
TS42446-0010EA 102 USD InStock
TS42446-0010 TS42446-0051
Sodium thiosulphate pentahydrate 99.5+% ACS
Sodium thiosulphate pentahydrate


Formula: Na₂S₂O₃.5H₂O
Melting Pt: 48.5 °C
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844
Order Now
Specification Test Results
Appearance White Crystals
Titration Iodimetric 99.5 to 101.0 %
pH 6.0 to 8.4 (5 % solution at 25.0°C)
Sulfide (S) Passes test (limit about 1 ppm)
SO4 and SO3 (as SO4) ≤0.1 %
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.002 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty