Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium chloride hexahydrate 98+% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC38926-0010 AC38926-0025 AC38926-2500
TS38926-0010EA 60 USD InStock
TS38926-0010 TS38926-0025 TS38926-2500
Calcium chloride hexahydrate 98+% for analysis
Calcium chloride hexahydrate


Formula: CaCl₂·6H₂O
MW: 219.08
Melting Pt: 29.54
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00149616
CAS Number: 7774-34-7
EINECS: 233-140-8
Merck Index: 13,01660
Order Now
Specification Test Results
Appearance Colorless Chunks
Titration Complexometric ≥98.0 %
pH 4.5 to 8.5 (5 % solution)
Phosphate (PO4) ≤0.003 %
Sulfate (SO4) ≤0.05 %
Ammonium (NH4) ≤0.01 %
Arsenic (As) ≤1 ppm
Copper (Cu) ≤0.002 %
Iron (Fe) ≤0.005 %
Lead (Pb) ≤0.002 %
Nickel (Ni) ≤0.002 %
Insoluble matter ≤0.025 % (in water and ppt by NH4OH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty