Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium chloride hexahydrate 99%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC21747-0010 AC21747-0025 AC21747-0100 AC21747-5000
TS21747-0010EA 137 USD InStock
TS21747-0010 TS21747-0025 TS21747-0100 TS21747-5000
Aluminium chloride hexahydrate 99%, pure
Aluminium chloride hexahydrate


Formula: AlCl₃·6H₂O
Density: 2.39 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
Order Now
Specification Test Results
Appearance White to yellow Adhering powder or crystals
Titration Argentometric ≥98.5 %
Water 42 to 48 % (K.F.)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty