Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium acetate 98%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC16988-0010 AC16988-0025 AC16988-0100
TS16988-0010EA 93 USD InStock
TS16988-0010 TS16988-0025 TS16988-0100
Ammonium acetate 98%, pure
Ammonium acetate


Formula: H₃CCOONH₄
Melting Pt: 114 °C
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=7903b63bd52460b98c28450c41ffbd0dbfc3377d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Adhering crystals
Infrared spectrum Conforms
Water ≤2 %
Titration with HClO4 ≥97.5 % (On anhydrous substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty