Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iodoethane 98% stabilised, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

Stabilization: Stabilised with silver

AC33212-0025 AC33212-1000 AC33212-5000
TS33212-0025EA 630 USD InStock
TS33212-0025 TS33212-1000 TS33212-5000
Iodoethane 98% stabilised, pure
Iodoethane


Formula: C₂H₅I
MW: 155.97 g/mol
Boiling Pt: 72
Melting Pt: -108
Density: 1.95 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00001091
CAS Number: 75-03-6
EINECS: 200-833-1
Merck Index: 13,03846
Order Now
Specification Test Results
Appearance Clear colorless to light yellow to pink Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer silver

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty