Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium cyanide 98+%, Extra Pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC37031-0010 AC37031-0050
TS37031-0010EA 161 USD InStock
TS37031-0010 TS37031-0050
Sodium cyanide 98+%, Extra Pure
Sodium cyanide


Formula: CNNa
Boiling Pt: 1496 °C (1013 hPa)
Melting Pt: 562 °C
Storage Temperature: Ambient
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1,I
Merck Index: 13,08679
Order Now
Specification Test Results
Appearance White Powder or granular powder or crystals
Titration Argentometric ≥98.0 %
Water ≤0.1 %
Sodium carbonate (Na2CO3) 0.2 to 0.8 %
Sodium hydroxide (NaOH) 0.3 to 0.8 %
Sodium formate 0.1 to 0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty