Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium carbonate 99.999% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC43680-0050 AC43680-0250
TS43680-0050EA 65 USD InStock
TS43680-0050 TS43680-0250
Sodium carbonate 99.999% (trace metals basis)
Sodium carbonate


Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
Merck Index: 14,08596
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=0a29c32fc35c3063a6cddfd3f5c9a58f119301fc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder
Total trace metal impurities ≤10 ppm

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty