Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rubidium chloride 99.8+% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC20126-0100 AC20126-0500 AC20126-2500
TS20126-0100EA 83 USD InStock
TS20126-0100 TS20126-0500 TS20126-2500
Rubidium chloride 99.8+% (trace metals basis)
Rubidium chloride


Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390
Melting Pt: 715
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9
Order Now
Specification Test Results
Appearance White Powder and/or chunks
Total trace metal impurities ≤2000.0 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty