Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium perchlorate 99+% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC19471-0050 AC19471-1000 AC19471-5000
TS19471-0050EA 54 USD InStock
TS19471-0050 TS19471-1000 TS19471-5000
Lithium perchlorate 99+% for analysis
Lithium perchlorate


Formula: ClLiO₄
MW: 106.39 g/mol
Boiling Pt: 430
Melting Pt: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD00011079
CAS Number: 7791-03-9
EINECS: 232-237-2
UN: 1481
ADR: 5.1 II
Merck Index: 13,05561
Order Now
Specification Test Results
Appearance White Crystalline or granular powder or crystals
Water ≤1.0 % (K.F.)
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤200 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤50 ppm
Potassium (K) ≤200 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty