Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium iodate 99+% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC20176-0025 AC20176-1000 AC20176-5000
TS20176-0025EA 732 USD InStock
TS20176-0025 TS20176-1000 TS20176-5000
Sodium iodate 99+% for analysis
Sodium iodate


Formula: INaO₃
MW: 197.89 g/mol
Melting Pt: 560 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1 II
Merck Index: 13,08704
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=806173453dcec1d2f791367d269d662d3459149a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Iodometric ≥99.0 %
Loss on drying ≤0.1 % (130°C)
Iodide (I) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤5 ppm
Nickel (Ni) ≤5 ppm
Nitrogen (N) ≤0.003 %
Potassium (K) ≤100 ppm
Impurity ≤0.02 % Bromide,bromate,chloride and chlorate (as Cl)
Insoluble matter ≤0.005 % (in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty