Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium nitrate, extra pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC43609-0010 AC43609-0050 AC43609-0250
TS43609-0010EA 60 USD InStock
TS43609-0010 TS43609-0050 TS43609-0250
Sodium nitrate, extra pure
Sodium nitrate


Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380
Melting Pt: 308
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
UN: 1498
ADR: 5.1 III
Merck Index: 13,08720
Order Now
Specification Test Results
Appearance Colorless to white Crystalline powder or crystals
Loss on drying ≤2 % (105°C)
pH 5.5 to 8.3 (5 % in water)
Titration after Ion exchange ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty