Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium superoxide

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC35542-0250 AC35542-5000
TS35542-0250EA 49 USD InStock
TS35542-0250 TS35542-5000
Potassium superoxide
Potassium superoxide


Formula: KO₂
Melting Pt: 400 °C
Storage Temperature: Ambient
MDL Number: MFCD00082411
CAS Number: 12030-88-5
EINECS: 234-746-5
UN: 2466
ADR: 5.1,I
Order Now
Specification Test Results
Appearance Yellow Powder or chunks
Oxygen (O) ≥215 cm³/g O2-content

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty