Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium cyanate 97%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC20111-0010 AC20111-1000 AC20111-5000
TS20111-0010EA 165 USD InStock
TS20111-0010 TS20111-1000 TS20111-5000
Potassium cyanate 97%, pure
Potassium cyanate


Formula: CKNO
Melting Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD00011411
CAS Number: 590-28-3
EINECS: 209-676-3
Merck Index: 12,07788
Order Now
Specification Test Results
Appearance White Crystalline powder and/or chunks
Titration Argentometric ≥96.0 %
Potassium carbonate ≤3 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty