Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perrhenic acid 99.99% (trace metals basis) in aqueous solution

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC31618-0050 AC31618-0250
TS31618-0050EA 142 USD InStock
TS31618-0050 TS31618-0250
Perrhenic acid 99.99% (trace metals basis) in aqueous solution
Perrhenic acid


Formula: HO₄Re
Storage Temperature: Ambient
MDL Number: MFCD00011326
CAS Number: 13768-11-1
EINECS: 237-380-4
UN: 3264
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Strength 70 to 76.5 % (HReO4)
Total trace metal impurities ≤100 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty