Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium chloride, anhydrous 99%, granules, extra pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC19578-0010 AC19578-5000
TS19578-0010EA 102 USD InStock
TS19578-0010 TS19578-5000
Aluminium chloride, anhydrous 99%, granules, extra pure
Aluminium chloride


Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Melting Pt: 194 °C
Density: 2.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335
Order Now
Specification Test Results
Appearance White to yellow to gray Granules
Titration Argentometric ≥98.5 %
Aluminium (Al) ≤150 ppm (free Al)
Chromium (Cr) ≤10 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤100 ppm
Nickel (Ni) ≤20 ppm
Zinc (Zn) ≤30 ppm
Insoluble matter ≤0.2 % (in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty