Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium tetrafluoroborate 99%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC31096-0025 AC31096-1000 AC31096-5000
TS31096-0025EA 339 USD InStock
TS31096-0025 TS31096-1000 TS31096-5000
Ammonium tetrafluoroborate 99%, pure
Ammonium tetrafluoroborate


Formula: H₄BF₄N
MW: 104.84
Melting Pt: 230 °C (subl.)
Density: 1.87
Storage Temperature: Ambient
MDL Number: MFCD06245785
CAS Number: 13826-83-0
EINECS: 237-531-4
UN: 3260
ADR: 8 III
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Titration with NaOH ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty