Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Antimony(III) bromide 97%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC31840-0500 AC31840-2500
TS31840-0500EA 123 USD InStock
TS31840-0500 TS31840-2500
Antimony(III) bromide 97%, pure
Antimony(III) bromide


Formula: Br₃Sb
Boiling Pt: 280 °C
Melting Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00016317
CAS Number: 7789-61-9
EINECS: 232-179-8
UN: 1549
ADR: 6.1,III
Merck Index: 12,00747
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Titration Argentometric ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty