Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium bromide, anhydrous 99%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC31538-0500 AC31538-2500
TS31538-0500EA 57 USD InStock
TS31538-0500 TS31538-2500
Barium bromide, anhydrous 99%, pure
Barium bromide


Formula: BaBr₂
MW: 297.14 g/mol
Boiling Pt: 1835
Melting Pt: 860
Storage Temperature: Ambient
MDL Number: MFCD00003444
CAS Number: 10553-31-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,00995
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Argentometric ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty