Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cadmium(II) carbonate, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC21232-0010 AC21232-2500
TS21232-0010EA 182 USD InStock
TS21232-0010 TS21232-2500
Cadmium(II) carbonate, pure
Cadmium(II) carbonate


Formula: CCdO₃
MW: 172.42 g/mol
Melting Pt: 357 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010918
CAS Number: 513-78-0
EINECS: 208-168-9
UN: 2570
ADR: 6.1 II
Merck Index: 12,01652
Order Now
Specification Test Results
Appearance White to off-white Powder
Assay 60.0 to 63.0 % (Cd)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty