Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indium(III) iodide 99.998% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC31601-0010 AC31601-0050 AC31601-0250
TS31601-0010EA 51 USD InStock
TS31601-0010 TS31601-0050 TS31601-0250
Indium(III) iodide 99.998% (trace metals basis)
Indium(III) iodide


Formula: I₃In
MW: 495.53 g/mol
Boiling Pt: 500
Melting Pt: 209.85
Storage Temperature: Ambient
MDL Number: MFCD00016151
CAS Number: 13510-35-5
EINECS: 236-839-6
Order Now
Specification Test Results
Appearance Yellow to light brown Powder or crystals
Titration Complexometric 22.7 to 23.5 % (In)
Total trace metal impurities ≤0.002 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty