Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Manganese(II) oxide 99.99% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC43697-0010 AC43697-0100
TS43697-0010EA 38 USD InStock
TS43697-0010 TS43697-0100
Manganese(II) oxide 99.99% (trace metals basis)
Manganese(II) oxide


Formula: MnO
MW: 70.94
Melting Pt: 1840
Density: 5.43
Storage Temperature: Ambient
MDL Number: MFCD00016226
CAS Number: 1344-43-0
Order Now
Specification Test Results
Appearance Green Crystalline powder and/or chunks
Titration Complexometric 76.7 to 78.2 % (Mn)
Total trace metal impurities ≤100 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty