Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyltriethoxysilane 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC17464-0010 AC17464-2500
TS17464-0010EA 148 USD InStock
TS17464-0010 TS17464-2500
Phenyltriethoxysilane 98%
Phenyltriethoxysilane


Formula: C₁₂H₂₀O₃Si
MW: 240.37 g/mol
Boiling Pt: 237
Melting Pt: 41
Density: 0.996 g/cm³
Flash Pt: 42 °C
MDL Number: MFCD00009065
CAS Number: 780-69-8
EINECS: 212-305-8
UN: 1993
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4594 to 1.4614 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty