Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium dihydrogen phosphate 98+% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC42338-0025 AC42338-5000
TS42338-0025EA 298 USD InStock
TS42338-0025 TS42338-5000
Ammonium dihydrogen phosphate 98+% ACS
Ammonium dihydrogen phosphate


Formula: NH₄H₂PO₄
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547
Order Now
Specification Test Results
Appearance White Crystalline solid
pH 3.8 to 4.4 (5 % at 25.0°C)
Titration with NaOH ≥98.0 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤0.001 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty