Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cadmium(II) sulphate 99+% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC27056-0025 AC27056-5000
TS27056-0025EA 807 USD InStock
TS27056-0025 TS27056-5000
Cadmium(II) sulphate 99+% for analysis
Cadmium(II) sulphate


Formula: CdO₄S
MW: 208.47
Melting Pt: 1000
Density: 4.7
Storage Temperature: Ambient
MDL Number: MFCD00010923
CAS Number: 10124-36-4
EINECS: 233-331-6
UN: 2570
ADR: 6.1 III
Merck Index: 13,01627
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Complexometric ≥99.0 %
pH 3.5 to 6.0 (5 % at 20°C)
Chloride (Cl) ≤5 ppm
Arsenic (As) ≤0.5 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤100 ppm
Zinc (Zn) ≤50 ppm
Alkalies+earths (Na,K,Ca) ≤0.05 %
Nitrogen compounds (as N) ≤0.004 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty