Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lead(II) sulphate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC38727-1000 AC38727-5000
TS38727-1000EA 40 USD InStock
TS38727-1000 TS38727-5000
Lead(II) sulphate 99%
Lead(II) sulphate


Formula: O₄PbS
MW: 303.26 g/mol
Melting Pt: 1000
Storage Temperature: Ambient
MDL Number: MFCD00011166
CAS Number: 7446-14-2
EINECS: 231-198-9
UN: 3077
ADR: 9 III
Merck Index: 12,05444
Order Now
Specification Test Results
Appearance White Crystalline powder
Identification Confirmed
Elemental Analysis ≥98.5 % (AES)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty