Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hydrogen sulphate 98.5% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC20775-0025 AC20775-5000
TS20775-0025EA 375 USD InStock
TS20775-0025 TS20775-5000
Potassium hydrogen sulphate 98.5% for analysis
Potassium hydrogen sulphate


Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695
Order Now
Specification Test Results
Appearance White Adhering crystals
Titration with NaOH 98.0 to 102.0 %
Chloride (Cl) ≤20 ppm
Ammonium (NH4) ≤20 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty