Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropylchlorosilane 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC37735-0050 AC37735-0250 AC37735-1000
TS37735-0050EA 56 USD InStock
TS37735-0050 TS37735-0250 TS37735-1000
Triisopropylchlorosilane 95%
Triisopropylchlorosilane


Formula: C₉H₂₁ClSi
Boiling Pt: 198 °C (760 mmHg)
Density: 0.902 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009656
CAS Number: 13154-24-0
UN: 2987
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty