Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulphuryl chloride 98.5%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC16945-0010 AC16945-0025
TS16945-0010EA 131 USD InStock
TS16945-0010 TS16945-0025
Sulphuryl chloride 98.5%
Sulphuryl chloride


Formula: Cl₂O₂S
MW: 134.97 g/mol
Boiling Pt: 69.1
Melting Pt: -54
Density: 1.667 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011451
CAS Number: 7791-25-5
EINECS: 232-245-6
UN: 1834
ADR: 6.1 I
Merck Index: 12,09153
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Titration Argentometric ≥98.0 % (ex Cl)
Free chlorine ≤0.1 %
Sulfur dioxide (SO2) ≤0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty