Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium oxalate ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC44696-1000 AC44696-5000
TS44696-1000EA 60 USD InStock
TS44696-1000 TS44696-5000
Disodium oxalate ACS
Disodium oxalate


Formula: Na₂C₂O₄
MW: 134 g/mol
Melting Pt: 250…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
Merck Index: 13,08723
Order Now
Specification Test Results
Appearance White Crystalline powder
Loss on drying ≤0.01 %
Titration with KMnO4 ≥99.5 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤0.002 %
Sulfate (SO4) ≤0.002 %
Ammonium (NH4) ≤0.002 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.005 %
Neutrality Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty