Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tributyltin hydride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC21573-0100 AC21573-0500 AC21573-2500
TS21573-0100EA 45 USD InStock
TS21573-0100 TS21573-0500 TS21573-2500
Tributyltin hydride 97%
Tributyltin hydride


Formula: C₁₂H₂₈Sn
Storage Temperature: Refrigerator
MDL Number: MFCD00009416
CAS Number: 688-73-3
EINECS: 211-704-4
UN: 2788
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4740 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty