Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cobalt(II) sulphate heptahydrate 99+%, extra pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC21310-0025 AC21310-1000 AC21310-5000
TS21310-0025EA 413 USD InStock
TS21310-0025 TS21310-1000 TS21310-5000
Cobalt(II) sulphate heptahydrate 99+%, extra pure
Cobalt(II) sulphate heptahydrate


Formula: CoSO₄·7H₂O
MW: 281.1 g/mol
Melting Pt: 735
Storage Temperature: Ambient
MDL Number: MFCD00149658
CAS Number: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9 III
Merck Index: 12,02510
Order Now
Specification Test Results
Appearance Red-brown Adhering crystalline powder or crystals
Titration Complexometric ≥99.0 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤500 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty