Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyltrimethoxysilane 97%, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC43308-0010 AC43308-1000
TS43308-0010EA 210 USD InStock
TS43308-0010 TS43308-1000
Ethyltrimethoxysilane 97%, AcroSeal™
Ethyltrimethoxysilane


Formula: C₅H₁₄O₃Si
MW: 150.25 g/mol
Boiling Pt: 123…124 °C (1013 hPa)
Density: 0.94 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00051541
CAS Number: 5314-55-6
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty