Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Digadolinium trioxide 99.9% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC31551-0100 AC31551-0250 AC31551-1000
TS31551-0100EA 43 USD InStock
TS31551-0100 TS31551-0250 TS31551-1000
Digadolinium trioxide 99.9% (trace metals basis)
Digadolinium trioxide


Formula: Gd₂O₃
Melting Pt: 1900 °C
Storage Temperature: Ambient
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9
Order Now
Specification Test Results
Appearance White Powder
Total conc. of imp.as REO ≤0.1 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty