Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyltriethoxysilane 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC17462-0010 AC17462-2500
TS17462-0010EA 157 USD InStock
TS17462-0010 TS17462-2500
Methyltriethoxysilane 98+%
Methyltriethoxysilane


Formula: C₇H₁₈O₃Si
MW: 178.3 g/mol
Boiling Pt: 141…143 °C (760 mmHg)
Melting Pt: -50
Density: 0.89 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009066
CAS Number: 2031-67-6
EINECS: 217-983-9
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3825 to 1.3845 (20°C, 589 nm)
GC ≥98.0 %
Hydrolyzable chloride ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty