Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel sulphate heptahydrate DIN 50970 for analysis, for nickel plating

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC27055-0010 AC27055-2500
TS27055-0010EA 267 USD InStock
TS27055-0010 TS27055-2500
Nickel sulphate heptahydrate DIN 50970 for analysis, for nickel plating
Nickel sulphate heptahydrate


Formula: NiO₄S·7H₂O
MW: 280.86
Melting Pt: 99
Density: 2
Storage Temperature: Ambient
MDL Number: MFCD00149814
CAS Number: 10101-98-1
EINECS: 232-104-9
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance Green to dark green Powder or crystals or needles
Titration Complexometric 20 to 21 % (Ni)
Arsenic (As) ≤10 ppm
Cadmium (Cd) ≤10 ppm
Cobalt (Co) ≤5000 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤20 ppm
Lead (Pb) ≤10 ppm
Zinc (Zn) ≤30 ppm
Insoluble matter ≤0.01 % (in 0.1 N H2SO4)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty