Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Platinum 5% on activated carbon hydrogenation catalyst

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC19523-0100 AC19523-0500
TS19523-0100EA 149 USD InStock
TS19523-0100 TS19523-0500
Platinum 5% on activated carbon hydrogenation catalyst
Platinum


Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1,III
Order Now
Specification Test Results
Appearance Black Powder
Assay 5 % (Pt)
Additional info < 85 um: 90%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty