Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropylsilane 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC21492-0100 AC21492-0500 AC21492-2500
TS21492-0100EA 55 USD InStock
TS21492-0100 TS21492-0500 TS21492-2500
Triisopropylsilane 98%
Triisopropylsilane


Formula: C₉H₂₂Si
MW: 158.36 g/mol
Boiling Pt: 84
Density: 0.773 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00009657
CAS Number: 6485-79-6
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4360 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty