Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vanadium(V) oxide 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC45016-0010 AC45016-0250 AC45016-2500
TS45016-0010EA 176 USD InStock
TS45016-0010 TS45016-0250 TS45016-2500
Vanadium(V) oxide 99+%
Vanadium(V) oxide


Formula: O₅V₂
MW: 181.88 g/mol
Boiling Pt: 1750
Melting Pt: 690
Density: 3.35 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011457
CAS Number: 1314-62-1
UN: 2862
ADR: 6.1 III
Merck Index: 13,09987
Order Now
Specification Test Results
Appearance Dark yellow Powder
Assay ≥99.0 %
Particle size ≤1.2 mm
Chromium (Cr) ≤0.1 %
Iron (Fe) ≤0.1 %
Phosphorus (P) ≤0.04 %
Potassium (K) ≤0.4 %
Silicon (Si) ≤0.06 %
Sodium (Na) ≤0.3 %
Sulfur (S) ≤0.07 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty