Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc iodide 98+%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC20806-0010 AC20806-0500 AC20806-2500
TS20806-0010EA 525 USD InStock
TS20806-0010 TS20806-0500 TS20806-2500
Zinc iodide 98+%, pure
Zinc iodide


Formula: I₂Zn
MW: 319.2 g/mol
Boiling Pt: 620
Melting Pt: 446
Density: 4.74 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8 II
Merck Index: 12,10269
Order Now
Specification Test Results
Appearance White to yellow to light beige Hygroscopic crystalline powder or crystals
Titration Argentometric ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty