Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin(II) fluoride 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC30850-0050 AC30850-1000 AC30850-5000
TS30850-0050EA 33 USD InStock
TS30850-0050 TS30850-1000 TS30850-5000
Tin(II) fluoride 99%
Tin(II) fluoride


Formula: SnF₂
MW: 156.71
Boiling Pt: 850
Melting Pt: 215
Density: 4.57
Storage Temperature: Ambient
MDL Number: MFCD00042540
CAS Number: 7783-47-3
EINECS: 231-999-3
UN: 2923
ADR: 8 III
Merck Index: 12,08940
Order Now
Specification Test Results
Appearance White to off-white Powder or crystals
Gravimetry ≥98.5 % (ex Sn)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty