Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodium 5% on activated carbon, dry, powder, unreduced

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC19544-0010 AC19544-0050 AC19544-0250
TS19544-0010EA 132 USD InStock
TS19544-0010 TS19544-0050 TS19544-0250
Rhodium 5% on activated carbon, dry, powder, unreduced
Rhodium


Formula: Rh
Boiling Pt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960 °C
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
Order Now
Specification Test Results
Appearance Gray-black to black Powder
Assay 4.5 to 5.5 % (Rh)
Particle Size/Sieve analysis <128 um: 99%
Additional info D50: 23 um

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty